Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Myb/SANT DNA binding domain containing 1 | MSANTD1 | C4orf44 | -128.5kb | Sense | Chr. 4 - 3243728 - 3256615 on NCBI Build GRCh38 |
Downstream | HGF activator | HGFAC | HGFA | 47.1kb | Sense | Chr. 4 - 3441933 - 3449495 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |