Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
mucin 12, cell surface associated |
MUC12 |
MUC-11, MUC-12, MUC11 |
Sense | Chr. 7 - 100969623 - 101018949 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC102724094 | LOC102724094 | 43.8kb | Anti-sense | Chr. 7 - 101014320 - 101017608 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |