Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mediator complex subunit 30 | MED30 |
MED30S, THRAP6, TRAP25 |
-482.1kb | Anti-sense | Chr. 8 - 117520546 - 117540262 on NCBI Build GRCh38 |
Downstream | sterile alpha motif domain containing 12 | SAMD12 | 106.9kb | Sense | Chr. 8 - 118131828 - 118621995 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |