Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chloride intracellular channel 5 | CLIC5 |
DFNB102, DFNB103, MST130, MSTP130 |
Sense | Chr. 6 - 45898451 - 46129809 on NCBI Build GRCh38 | |
Downstream | ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative) | ENPP4 | NPP4 | 201.3kb | Anti-sense | Chr. 6 - 46129964 - 46146699 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |