Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | N-acylethanolamine acid amidase | NAAA |
ASAHL, PLT |
Sense | Chr. 4 - 75910655 - 75941013 on NCBI Build GRCh38 | |
Downstream | SDA1 domain containing 1 | SDAD1 | 33.5kb | Sense | Chr. 4 - 75949906 - 75990962 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |