Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928471 | LOC101928471 | -13.6kb | Anti-sense | Chr. 12 - 24223271 - 24237965 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 477 | LINC00477 |
C12orf67, FAM191B |
313.4kb | Sense | Chr. 12 - 24566964 - 24584168 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |