Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110777.1 | 2 - 3 | 502 | 94 | ||
GenBank mRNA | AK056572.1 | - | 2 - 3 | 502 | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NSF attachment protein gamma | NAPG | GAMMASNAP | -110.6kb | Sense | Chr. 18 - 10525876 - 10552769 on NCBI Build GRCh38 |
Downstream | piezo type mechanosensitive ion channel component 2 | PIEZO2 |
C18orf30, C18orf58, DA3, DA5, FAM38B, FAM38B2, HsT748, HsT771, MWKS |
3.2kb | Anti-sense | Chr. 18 - 10670237 - 11149561 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |