Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MOB kinase activator 1A | MOB1A |
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B |
-17.8kb | Sense | Chr. 2 - 74152598 - 74178967 on NCBI Build GRCh38 |
Downstream |
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase |
MTHFD2 | NMDMC | 3.0 | Anti-sense | Chr. 2 - 74198563 - 74215298 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |