Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ankyrin repeat and kinase domain containing 1 | ANKK1 | PKK2 | -5.3kb | Anti-sense | Chr. 11 - 113386014 - 113400418 on NCBI Build GRCh38 |
Downstream | dopamine receptor D2 | DRD2 |
D2DR, D2R |
1.8kb | Sense | Chr. 11 - 113409595 - 113475279 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |