Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ribosomal protein S2 pseudogene 46 | RPS2P46 | RPS2_22_1525 | -85.8kb | Sense | Chr. 17 - 19445929 - 19446875 on NCBI Build GRCh38 |
Downstream |
small nucleolar RNA, H/ACA box 59B |
SNORA59B | ACA59 | 22.9kb | Anti-sense | Chr. 17 - 19557560 - 19557711 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |