Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ZFP91-CNTF readthrough (NMD candidate) | ZFP91-CNTF | Sense | Chr. 11 - 58579114 - 58625733 on NCBI Build GRCh38 | ||
Downstream | ciliary neurotrophic factor | CNTF | HCNTF | 31.1kb | Sense | Chr. 11 - 58622673 - 58625733 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |