Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CELF2 antisense RNA 1 | CELF2-AS1 | C10orf31 | -103.7kb | Anti-sense | Chr. 10 - 11316833 - 11344674 on NCBI Build GRCh38 |
Downstream | USP6 N-terminal like | USP6NL |
RNTRE, TRE2NL, USP6NL-IT1 |
8.2kb | Anti-sense | Chr. 10 - 11460510 - 11611789 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |