Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
ENST00000314018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
ENST00000314018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
ENST00000314018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
ENST00000609256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
ENST00000609256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
ENST00000609256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
ENST00000609256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
ENST00000609256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
NM_177455.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
NM_177455.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008454.hg.1 |
NM_177455.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
ENST00000314018.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
ENST00000314018.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
ENST00000314018.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
ENST00000609256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
ENST00000609256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
ENST00000609256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
ENST00000609256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
ENST00000609256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
NM_177455.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
NM_177455.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008454.hg.1 |
NM_177455.3 |
Refseq |