Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | matrix metallopeptidase 28 | MMP28 |
EPILYSIN, MM28, MMP-25, MMP-28, MMP25 |
-33.7kb | Anti-sense | Chr. 17 - 35756249 - 35795707 on NCBI Build GRCh38 |
Downstream | HEAT repeat containing 9 | HEATR9 | C17orf66 | 24.8kb | Anti-sense | Chr. 17 - 35854946 - 35868891 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |