Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK023731.1 | - | 1 | 744 | 76 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927342 | LOC101927342 | -150.7kb | Sense | Chr. 1 - 76011324 - 76019356 on NCBI Build GRCh38 | |
Downstream | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5 | ST6GALNAC5 |
SIAT7-E, SIAT7E, ST6GalNAcV |
695.6kb | Sense | Chr. 1 - 76867408 - 77069167 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |