Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PC4 and SFRS1 interacting protein 1 | PSIP1 |
DFS70, LEDGF, PAIP, PSIP2, p52, p75 |
Anti-sense | Chr. 9 - 15464066 - 15511019 on NCBI Build GRCh38 | |
Downstream | coiled-coil domain containing 171 | CCDC171 |
C9orf93, bA536D16.1, bA778P13.1 |
84.7kb | Sense | Chr. 9 - 15552874 - 16062431 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |