Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ankyrin repeat and SOCS box containing 13 | ASB13 | -39.4kb | Anti-sense | Chr. 10 - 5638857 - 5666595 on NCBI Build GRCh38 | |
Downstream | GDP dissociation inhibitor 2 | GDI2 |
HEL-S-46e, RABGDIB |
58.6kb | Anti-sense | Chr. 10 - 5765222 - 5813549 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |