Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TBC1 domain family member 8 | TBC1D8 |
AD3, GRAMD8, HBLP1, TBC1D8A, VRP |
-5.5kb | Sense | Chr. 2 - 101006435 - 101195326 on NCBI Build GRCh38 |
Downstream | CCR4-NOT transcription complex subunit 11 | CNOT11 |
C2orf29, C40 |
49.3kb | Anti-sense | Chr. 2 - 101252883 - 101270316 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |