Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | telomeric repeat binding factor 1 | TERF1 |
PIN2, TRBF1, TRF, TRF1, hTRF1-AS, t-TRF1 |
-4.1kb | Sense | Chr. 8 - 73008862 - 73047752 on NCBI Build GRCh38 |
Downstream | somatomedin B and thrombospondin type 1 domain containing | SBSPON |
C8orf84, RPESP |
8.9kb | Anti-sense | Chr. 8 - 73063077 - 73093272 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |