Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK128255.1 | - | 2 | 1655 | 98 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hydroxyacyl-CoA dehydrogenase | HADH |
HAD, HADH1, HADHSC, HCDH, HHF4, MSCHAD, SCHAD |
-12.4kb | Anti-sense | Chr. 4 - 107989714 - 108035175 on NCBI Build GRCh38 |
Downstream | LEF1 antisense RNA 1 | LEF1-AS1 | LEF1NAT | 117.3kb | Anti-sense | Chr. 4 - 108167525 - 108176430 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |