Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
URI1, prefoldin like chaperone |
URI1 |
C19orf2, NNX3, PPP1R19, RMP, URI |
Sense | Chr. 19 - 29923644 - 30016612 on NCBI Build GRCh38 | |
Downstream | zinc finger protein 536 | ZNF536 | 299.2kb | Sense | Chr. 19 - 30224236 - 30713538 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |