Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322030.1 | NP_001308959.1 | 10 | 1661 | 105 | |
XM_017021429.1 | XP_016876918.1 | 11 | 1861 | 105 | ||
XM_017021430.1 | XP_016876919.1 | 11 | 1846 | 105 | ||
GenBank mRNA | BC036547.1 | - | 1 | 1854 | 105 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | intraflagellar transport 43 | IFT43 |
C14orf179, CED3 |
-112.0kb | Sense | Chr. 14 - 75985753 - 76084073 on NCBI Build GRCh38 |
Downstream | estrogen related receptor beta | ESRRB |
DFNB35, ERR2, ERRb, ESRL2, NR3B2 |
171.8kb | Sense | Chr. 14 - 76371347 - 76501839 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |