Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120653.1 | 3 | 852 | 102 | ||
GenBank mRNA | BC043271.2 | - | 3 | 852 | 102 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | KIAA1462 | KIAA1462 | JCAD | -115.4kb | Anti-sense | Chr. 10 - 30012800 - 30069816 on NCBI Build GRCh38 |
Downstream | mitochondrial poly(A) polymerase | MTPAP |
PAPD1, SPAX4 |
77.1kb | Anti-sense | Chr. 10 - 30309801 - 30349338 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |