Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TraB domain containing 2A | TRABD2A |
C2orf89, TIKI1 |
Anti-sense | Chr. 2 - 84821650 - 84881975 on NCBI Build GRCh38 | |
Downstream | thymosin beta 10 | TMSB10 |
MIG12, TB10 |
68.1kb | Sense | Chr. 2 - 84905639 - 84906675 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |