Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105372068 | LOC105372068 | -138.8kb | Anti-sense | Chr. 18 - 37274460 - 37276400 on NCBI Build GRCh38 | |
Downstream | microRNA 4318 | MIR4318 | 240.9kb | Anti-sense | Chr. 18 - 37657135 - 37657215 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |