Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | trans-L-3-hydroxyproline dehydratase | L3HYPDH | C14orf149 | -8.4kb | Sense | Chr. 14 - 59465545 - 59484438 on NCBI Build GRCh38 |
Downstream | coiled-coil domain containing 175 | CCDC175 |
C14orf38, c14_5395 |
10.9kb | Sense | Chr. 14 - 59504212 - 59577276 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |