Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000600.4 | NP_000591.1 | 4 | 562 | 146 | |
NM_001318095.1 | NP_001305024.1 | 3 | 371 | 146 | ||
XM_011515390.2 | XP_011513692.1 | 5 | 895 | 146 | ||
GenBank mRNA | A09363.1 | - | 4 | 146 | ||
BC015511.1 | - | 4 | 490 | 3884652 | 146 | |
BT019748.1 | - | 4 | 441 | 146 | ||
BT019749.1 | - | 4 | 441 | 146 | ||
CD013918.1 | - | 3 | 299 | 146 | ||
CR450296.1 | - | 4 | 441 | 146 | ||
M14584.1 | - | 4 | 146 | |||
M18403.1 | - | 4 | 503 | 146 | ||
M29150.1 | - | 4 | 476 | 146 | ||
M54894.1 | - | 4 | 491 | 146 | ||
S56892.1 | - | 4 | 456 | 146 | ||
X04403.1 | - | 3 | 348 | 146 | ||
X04430.1 | - | 4 | 146 | |||
X04602.1 | - | 4 | 474 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Homo sapiens Inventoried Amplicon spans exons and probe does not span exons |