Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020926.hg.1 |
ENST00000301691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020926.hg.1 |
NM_025237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020926.hg.1 |
TC17001562.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010797.hg.1 |
ENST00000301691.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010797.hg.1 |
ENST00000301691.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010797.hg.1 |
ENST00000301691.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010797.hg.1 |
ENST00000301691.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010797.hg.1 |
NM_025237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010797.hg.1 |
NM_025237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010797.hg.1 |
NM_025237.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010797.hg.1 |
NM_025237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186088.hg.1 |
ENST00000301691.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186088.hg.1 |
NM_025237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186088.hg.1 |
ENST00000301691.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186088.hg.1 |
NM_025237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186088.hg.1 |
TC1700010797.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010797.hg.1 |
ENST00000301691.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010797.hg.1 |
ENST00000301691.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010797.hg.1 |
ENST00000301691.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010797.hg.1 |
ENST00000301691.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010797.hg.1 |
NM_025237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010797.hg.1 |
NM_025237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010797.hg.1 |
NM_025237.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010797.hg.1 |
NM_025237.2 |
Refseq |