Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036299.hg.1 |
TC01002352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036299.hg.1 |
ENST00000374475.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036299.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036299.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036294.hg.1 |
TC01002352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036294.hg.1 |
ENST00000374475.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036294.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036294.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036298.hg.1 |
TC01002352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036298.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036298.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036297.hg.1 |
TC01002352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036297.hg.1 |
ENST00000374475.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036297.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036297.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036303.hg.1 |
TC01002352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036303.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198733.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198733.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198733.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198733.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198733.hg.1 |
TC0100013305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198732.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198732.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198732.hg.1 |
ENST00000374479.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198732.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198732.hg.1 |
TC0100013305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100198736.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198736.hg.1 |
NM_000147.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198736.hg.1 |
TC0100013305.hg.1 |
Clariom D TC |