Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019860.hg.1 |
TC07001276.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019860.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019860.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019860.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011069.hg.1 |
ENST00000541353.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011069.hg.1 |
ENST00000541353.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011069.hg.1 |
ENST00000541353.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011069.hg.1 |
ENST00000541353.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011069.hg.1 |
ENST00000541353.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011069.hg.1 |
ENST00000543539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011069.hg.1 |
ENST00000543539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011069.hg.1 |
ENST00000543539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011069.hg.1 |
ENST00000543539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700176371.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700176371.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700176371.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700176371.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700176371.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700176371.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700176371.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700176371.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700176371.hg.1 |
TC0700010719.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011069.hg.1 |
ENST00000541353.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011069.hg.1 |
ENST00000541353.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011069.hg.1 |
ENST00000541353.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011069.hg.1 |
ENST00000541353.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011069.hg.1 |
ENST00000541353.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011069.hg.1 |
ENST00000543539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011069.hg.1 |
ENST00000543539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011069.hg.1 |
ENST00000543539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011069.hg.1 |
ENST00000543539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000408931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
ENST00000492961.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001077653.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010719.hg.1 |
NM_001166220.1 |
Refseq |