Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165947.2 | NP_001159419.1 | 1 - 2 | 408 | 93 | |
GenBank mRNA | AK302787.1 | - | 1 - 2 | 247 | 93 | |
BX647777.1 | - | 1 - 2 | 192 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Homo sapiens Probe spans exons |