Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014829.hg.1 |
ENST00000317814.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014829.hg.1 |
ENST00000541682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014829.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014829.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014829.hg.1 |
TC17001103.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175218.hg.1 |
TC1700009671.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000317814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000541682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
ENST00000577735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_001165967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009671.hg.1 |
NM_032580.3 |
Refseq |