Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000517440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000517440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000517440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000517440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000517440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028470.hg.1 |
TC05002071.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028470.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028470.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028470.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028470.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189393.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189393.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189393.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189393.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189393.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189393.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189393.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189393.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189393.hg.1 |
TC0500012864.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000329198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000517440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000517440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000517440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000517440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000517440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012864.hg.1 |
NM_004387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189392.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189392.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189392.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189392.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189392.hg.1 |
ENST00000424406.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189392.hg.1 |
ENST00000521848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189392.hg.1 |
NM_001166175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189392.hg.1 |
NM_001166176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189392.hg.1 |
TC0500012864.hg.1 |
Clariom D TC |