Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000163.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000163.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000163.hg.1 |
TC14000039.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006492.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006492.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006492.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006492.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006492.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006492.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006492.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006492.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006492.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006492.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006492.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006492.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144319.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144319.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144319.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144319.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144319.hg.1 |
TC1400006492.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400144318.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144318.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144318.hg.1 |
ENST00000285600.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144318.hg.1 |
NM_001004063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144318.hg.1 |
TC1400006492.hg.1 |
Clariom D TC |