Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs04176513_m1
See other TMCC1 GE Assays ›
Gene Symbol:
TMCC1
Gene Name:
transmembrane and coiled-coil domain family 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 129647792 - 129893605 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04176513_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TMCC1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001017395.3
NP_001017395.2
4 - 5
1943
91
NM_001128224.2
NP_001121696.1
4 - 5
1344
91
NR_033361.1
2 - 3
1489
91
XM_006713542.3
XP_006713605.1
5 - 6
2056
91
XM_006713543.3
XP_006713606.1
5 - 6
2053
91
XM_006713544.1
XP_006713607.1
5 - 6
2181
91
XM_006713550.3
XP_006713613.1
5 - 6
1614
91
XM_006713552.3
XP_006713615.1
2 - 3
1236
91
XM_006713553.3
XP_006713616.1
2 - 3
2528
91
XM_011512572.2
XP_011510874.1
6 - 7
2216
91
XM_011512573.2
XP_011510875.1
5 - 6
2795
91
XM_011512576.2
XP_011510878.1
5 - 6
2049
91
XM_011512579.2
XP_011510881.1
4 - 5
1804
91
XM_011512580.2
XP_011510882.1
7 - 8
1941
91
XM_011512581.2
XP_011510883.1
6 - 7
1778
91
XM_011512582.1
XP_011510884.1
2 - 3
1570
91
XM_011512583.2
XP_011510885.1
4 - 5
1347
91
XM_011512584.2
XP_011510886.1
6 - 7
1666
91
XM_017005931.1
XP_016861420.1
5 - 6
1964
91
XM_017005932.1
XP_016861421.1
6 - 7
2203
91
XM_017005933.1
XP_016861422.1
4 - 5
2000
91
XM_017005934.1
XP_016861423.1
8 - 9
2014
91
XM_017005935.1
XP_016861424.1
9 - 10
2103
91
XM_017005936.1
XP_016861425.1
6 - 7
1777
91
XM_017005937.1
XP_016861426.1
6 - 7
1711
91
XM_017005938.1
XP_016861427.1
8 - 9
2077
91
XM_017005939.1
XP_016861428.1
7 - 8
1971
91
XM_017005940.1
XP_016861429.1
4 - 5
1349
91
XM_017005941.1
XP_016861430.1
4 - 5
1475
91
GenBank mRNA
AB018322.1
-
1 - 2
510
91
AK124671.1
-
2 - 3
1490
91
AK291448.1
-
4 - 5
1762
91
AK293408.1
-
2 - 3
1026
91
BC012480.1
-
1 - 2
73
91
BC030619.2
-
2 - 3
1407
91
BC034284.1
-
1 - 2
93
91
BC039859.1
-
4 - 5
1850
5527623
91
BC047517.1
-
2 - 3
1407
91
BX640712.1
-
2 - 3
997
91
CR749206.1
-
2 - 3
1469
91
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene TMCC1
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0