Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VANGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172411.1 | NP_001165882.1 | 4 - 5 | 1074 | 105 | |
NM_001172412.1 | NP_001165883.1 | 4 - 5 | 1024 | 105 | ||
NM_138959.2 | NP_620409.1 | 4 - 5 | 1080 | 105 | ||
GenBank mRNA | AB057596.1 | - | 4 - 5 | 1024 | 105 | |
AB075805.1 | - | 3 - 4 | 821 | 105 | ||
AF481859.1 | - | 4 - 5 | 1036 | 105 | ||
BC032773.1 | - | 2 - 3 | 748 | 105 | ||
BC065272.1 | - | 4 - 5 | 1057 | 6165098 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |