Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046308.hg.1 |
TC02002879.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046308.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046308.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046308.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046308.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046308.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046311.hg.1 |
TC02002879.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046311.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046311.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046311.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046311.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046311.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000427571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000427571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000427571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000438786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000438786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000438786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000438786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000444142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000444142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
ENST00000444142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046309.hg.1 |
TC02002879.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046309.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046309.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046309.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046309.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046309.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000427571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000427571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000427571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000438786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000438786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000438786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000438786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000444142.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000444142.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
ENST00000444142.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016073.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
ENST00000444142.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
ENST00000444142.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218061.hg.1 |
TC0200016073.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
ENST00000438786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
ENST00000438786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
NM_001172417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218065.hg.1 |
TC0200016073.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
ENST00000233826.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
ENST00000409779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
ENST00000410029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
NM_001172416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
NM_002242.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200218062.hg.1 |
TC0200016073.hg.1 |
Clariom D TC |