Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014565.2 | NP_055380.2 | 1 | 742 | 74 | |
GenBank mRNA | BC069169.1 | - | 1 | 742 | 7216916 | 74 |
BC128248.1 | - | 1 | 742 | 74 | ||
BC141934.1 | - | 1 | 742 | 40114773 | 74 | |
BC141935.1 | - | 1 | 742 | 40114774 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |