Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020967.2 | NP_066018.1 | 5 | 723 | 68 | |
XM_005260474.3 | XP_005260531.1 | 4 | 677 | 68 | ||
XM_011528951.2 | XP_011527253.1 | 5 | 738 | 68 | ||
XM_017027988.1 | XP_016883477.1 | 4 | 677 | 68 | ||
GenBank mRNA | AB046857.1 | - | 3 | 271 | 68 | |
AF230533.1 | - | 5 | 695 | 68 | ||
AF470686.1 | - | 4 | 1099 | 68 | ||
BC140836.1 | - | 4 | 612 | 9021704 | 68 | |
BC151133.1 | - | 4 | 612 | 9057033 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |