Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031121.hg.1 |
TC01002031.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031121.hg.1 |
ENST00000359682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031121.hg.1 |
NM_001004688.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012348.hg.1 |
ENST00000366476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012348.hg.1 |
NM_001004690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012350.hg.1 |
ENST00000359682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012350.hg.1 |
NM_001004688.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012348.hg.1 |
ENST00000366476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012348.hg.1 |
NM_001004690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190622.hg.1 |
ENST00000359682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100190622.hg.1 |
NM_001004688.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190622.hg.1 |
ENST00000359682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100190622.hg.1 |
NM_001004688.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190622.hg.1 |
TC0100012350.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012350.hg.1 |
ENST00000359682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012350.hg.1 |
NM_001004688.1 |
Refseq |