Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007701.hg.1 |
ENST00000302622.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007701.hg.1 |
ENST00000623396.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007701.hg.1 |
ENST00000623976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007701.hg.1 |
NM_001005186.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005777.hg.1 |
ENST00000302622.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005777.hg.1 |
NM_001005186.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005777.hg.1 |
TC11000474.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100154264.hg.1 |
ENST00000302622.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154264.hg.1 |
ENST00000623396.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154264.hg.1 |
ENST00000623976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154264.hg.1 |
NM_001005186.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154264.hg.1 |
ENST00000302622.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154264.hg.1 |
ENST00000623396.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154264.hg.1 |
ENST00000623976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154264.hg.1 |
NM_001005186.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154264.hg.1 |
TC1100007701.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007701.hg.1 |
ENST00000302622.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007701.hg.1 |
ENST00000623396.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007701.hg.1 |
ENST00000623976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007701.hg.1 |
NM_001005186.2 |
Refseq |