Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005191.2 | NP_001005191.1 | 1 | 483 | 92 | |
GenBank mRNA | BC137147.1 | - | 1 | 483 | 9021144 | 92 |
BC137162.1 | - | 1 | 483 | 9021159 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |