Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010126.hg.1 |
ENST00000303532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010126.hg.1 |
NM_001005465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010126.hg.1 |
TC14000924.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008638.hg.1 |
ENST00000303532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008638.hg.1 |
NM_001005465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400162526.hg.1 |
ENST00000303532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400162526.hg.1 |
NM_001005465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400162526.hg.1 |
ENST00000303532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400162526.hg.1 |
NM_001005465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400162526.hg.1 |
TC1400008638.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400008638.hg.1 |
ENST00000303532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008638.hg.1 |
NM_001005465.1 |
Refseq |