Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039496.1 | NP_001034585.1 | 4 - 5 | 520 | 89 | |
GenBank mRNA | AL117564.1 | - | 4 - 5 | 523 | 89 |
Gene Symbol: KCNK4-TEX40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_133662.1 | 10 - 11 | 2507 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Homo sapiens 1700 Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28250712 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838428 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28250696 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838433 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28250713 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28250711 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838443 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838426 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28250716 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838436 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838429 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838424 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838431 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28250694 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838432 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838430 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07838435 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 65065 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 29919 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 116199 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 118766 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 250741 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P202773 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 8460 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 17599 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02457341 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P39381700 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P19262433 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P19262427 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P19262433 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P202773 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P202773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250700 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250711 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838431 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838428 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838430 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838426 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838433 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838435 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250713 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250716 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838443 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250716 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838433 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838426 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250713 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250711 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250700 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838443 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838435 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838428 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28250696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838430 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07838431 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P106991 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P202773 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 73315 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19262431 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19262427 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P106991 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P39381700 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838431 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838430 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838457 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460820 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838366 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460850 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460822 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460826 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838433 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838445 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838426 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838367 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460800 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838381 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838442 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838455 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460849 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460801 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838438 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460797 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838450 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460803 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838443 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838378 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838435 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838427 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838363 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838385 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460831 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838387 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838369 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460817 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838459 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460846 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838461 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16460844 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838448 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838464 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838424 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838440 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838451 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838467 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838454 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07838463 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P106991 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19262427 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001039496_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7940984 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7940983 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7940985 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7940986 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7940971 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 219883_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11724637_s_at | - | - |
Affymetrix Human Genome U219 Array | 11724637_PM_s_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR11007314.hg.1 | ENST00000328404.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11007314.hg.1 | ENST00000539086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11007314.hg.1 | ENST00000539852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11007314.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11007314.hg.1 | ENST00000544845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11007314.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11007314.hg.1 | TC11000594.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000535981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000535981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000535981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013034.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100201120.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100201120.hg.1 | ENST00000535981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100201120.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100201120.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100201120.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100201120.hg.1 | ENST00000535981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100201120.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100201120.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100201120.hg.1 | TC1100013034.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000328404.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000535981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000535981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000535981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | ENST00000539943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | NM_001039496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013034.hg.1 | NM_001039496.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 219883_at | - | - |
Human Genome U133 Plus 2.0 Array | 219883_PM_at | - | - |
Human Genome U133A 2.0 Array | 219883_PM_at | - | - |
Human Genome U133A 2.0 Array | 219883_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 219883_at6 | - | - |
Human Genome U95 Set | 47600_PM_at | - | - |
Human Genome U95 Set | 47600_at | - | - |
Human Genome U133 Set | 219883_at | - | - |
Human Genome U133 Set | 219883_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046551 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046551 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 118802 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3248086 | NM_001039496.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3248086 | AL117564.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3248086 | CR624550.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3248086 | NM_001039496.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3248086 | AL117564.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3248086 | CR624550.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248086 | NM_001039496.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248086 | AL117564.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248086 | CR624550.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248086 | NM_001039496.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248086 | AL117564.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248086 | CR624550.1 | human_transcriptome |