Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006474.hg.1 |
TC01000423.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006474.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006474.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007671.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153884.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153884.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153884.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153884.hg.1 |
ENST00000409358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153884.hg.1 |
ENST00000487056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153884.hg.1 |
NM_001099434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153884.hg.1 |
TC0100007671.hg.1 |
Clariom D TC |