Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008080.hg.1 |
ENST00000329196.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008080.hg.1 |
ENST00000329196.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008080.hg.1 |
ENST00000329196.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008080.hg.1 |
NM_175882.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008080.hg.1 |
NM_175882.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008080.hg.1 |
ENST00000329196.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008080.hg.1 |
ENST00000329196.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008080.hg.1 |
ENST00000329196.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008080.hg.1 |
NM_175882.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008080.hg.1 |
NM_175882.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159229.hg.1 |
ENST00000329196.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159229.hg.1 |
NM_175882.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159229.hg.1 |
ENST00000329196.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159229.hg.1 |
NM_175882.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159229.hg.1 |
TC1700008080.hg.1 |
Clariom D TC |