Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007526.hg.1 |
ENST00000335426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007526.hg.1 |
ENST00000423711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007526.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007526.hg.1 |
TC19000550.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000597876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000597876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000597876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000597876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000597876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000597876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008062.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
NM_182704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
TC1900008062.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
ENST00000335426.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
ENST00000423711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
ENST00000600586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
ENST00000622070.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160841.hg.1 |
NM_182704.1 |
Refseq |