Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
TC02000240.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
ENST00000234195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
ENST00000354545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
ENST00000406384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
ENST00000407257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
ENST00000417700.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
ENST00000469469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
NM_001170791.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
NM_001170792.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
NM_001170793.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003592.hg.1 |
NM_144713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000234195.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000354545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000406384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000407257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000417700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000425641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000442857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000469469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
NM_001170791.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
NM_001170792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
NM_001170793.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
NM_144713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000234195.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000354545.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000406384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000407257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000417700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000425641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000442857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
ENST00000469469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
NM_001170791.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
NM_001170792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
NM_001170793.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
NM_144713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150349.hg.1 |
TC0200007273.hg.1 |
Clariom D TC |