Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016365.hg.1 |
TC06001428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016365.hg.1 |
ENST00000355981.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016365.hg.1 |
NM_003546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011234.hg.1 |
ENST00000618305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011234.hg.1 |
NM_003546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011234.hg.1 |
NM_003546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011234.hg.1 |
ENST00000618305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011234.hg.1 |
NM_003546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011234.hg.1 |
NM_003546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175740.hg.1 |
ENST00000618305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175740.hg.1 |
NM_003546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175740.hg.1 |
ENST00000618305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175740.hg.1 |
NM_003546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175740.hg.1 |
TC0600011234.hg.1 |
Clariom D TC |