Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011376.hg.1 |
ENST00000371852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011376.hg.1 |
ENST00000371852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011376.hg.1 |
ENST00000371852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011376.hg.1 |
NM_003956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011376.hg.1 |
NM_003956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018962.hg.1 |
ENST00000371852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018962.hg.1 |
NM_003956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018962.hg.1 |
TC10001504.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000011376.hg.1 |
ENST00000371852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011376.hg.1 |
ENST00000371852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011376.hg.1 |
ENST00000371852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011376.hg.1 |
NM_003956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011376.hg.1 |
NM_003956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177270.hg.1 |
ENST00000371852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177270.hg.1 |
NM_003956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177270.hg.1 |
ENST00000371852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177270.hg.1 |
NM_003956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177270.hg.1 |
TC1000011376.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000177269.hg.1 |
ENST00000371852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177269.hg.1 |
NM_003956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177269.hg.1 |
ENST00000371852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177269.hg.1 |
NM_003956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177269.hg.1 |
TC1000011376.hg.1 |
Clariom D TC |